Product Details

SNP ID
rs28369552
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31400677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCACTTAGGGCTGCTTAAGGCT[C/G]GTCTTGCTAAGCTTCGTCGAGAACT
Phenotype
MIM: 603952
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DRG1 PubMed Links

Gene Details

Gene
DRG1
Gene Name
developmentally regulated GTP binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004147.3 Intron NP_004138.1

View Full Product Details