Product Details

SNP ID
rs35129608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:36863297 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAGATGGGACTGCATGACCACCC[A/G]GAGACATAAGTACTTCTCCTGTCCC
Phenotype
MIM: 601488
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NCF4 PubMed Links

Gene Details

Gene
NCF4
Gene Name
neutrophil cytosolic factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000631.4 Intron NP_000622.2
NM_013416.3 Intron NP_038202.2
XM_017028808.1 Intron XP_016884297.1

View Full Product Details