Product Details

SNP ID
rs28702416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:39702564 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACTGCACCTGGCCAACATTTTCT[A/C]GTCTCATAGGGCGTGTTTAAATCTA
Phenotype
MIM: 602846
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
UBE2K PubMed Links

Gene Details

Gene
UBE2K
Gene Name
ubiquitin conjugating enzyme E2 K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111112.1 Intron NP_001104582.1
NM_001111113.1 Intron NP_001104583.1
NM_001312646.1 Intron NP_001299575.1
NM_001312647.1 Intron NP_001299576.1
NM_001312648.1 Intron NP_001299577.1
NM_005339.4 Intron NP_005330.1

View Full Product Details