Product Details

SNP ID
rs28611147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:102944060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAATACACAGCCACAAGAAAGAA[C/T]GAGATCATGTCCTTTGGAGCAACGT
Phenotype
MIM: 611527
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC9B1 PubMed Links

Gene Details

Gene
SLC9B1
Gene Name
solute carrier family 9 member B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100874.2 Intron NP_001094344.1
NM_139173.3 Intron NP_631912.2
XM_006714093.3 Intron XP_006714156.1
XM_011531622.1 Intron XP_011529924.1
XM_011531623.1 Intron XP_011529925.1

View Full Product Details