Product Details

SNP ID
rs28695846
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:77862859 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGGAGTGCCCAACATGGCGGCGG[C/G]CGTAAGGTGCATGGGTAGAGGTAAG
Phenotype
MIM: 611821
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MRPL1 PubMed Links

Gene Details

Gene
MRPL1
Gene Name
mitochondrial ribosomal protein L1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020236.3 209 Missense Mutation GCC,GGC A4G NP_064621.3
XM_011532198.2 209 Missense Mutation GCC,GGC A4G XP_011530500.1
XM_011532199.1 209 Missense Mutation GCC,GGC A4G XP_011530501.1

View Full Product Details