Product Details
- SNP ID
-
rs28453128
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:218287757 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTTATTATATTTTAACATATCTC[C/T]CTCCTTATAGAGATAAGTTCCCCAA
- Phenotype
-
MIM: 611193
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RRP15
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116504418] are located under a probe and SNP(s) [rs35931083] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RRP15
- Gene Name
- ribosomal RNA processing 15 homolog
View Full Product Details