Product Details

SNP ID
rs17163414
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:222673007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAAGATGAAAACTGAATCATCA[A/G]GGAACCTTCACGCACTCCTGCTTCT
Phenotype
MIM: 612375 MIM: 613455
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AIDA PubMed Links

Gene Details

Gene
AIDA
Gene Name
axin interactor, dorsalization associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022831.2 Intron NP_073742.2
Gene
MIA3
Gene Name
MIA family member 3, ER export factor
There are no transcripts associated with this gene.

View Full Product Details