Product Details
- SNP ID
-
rs2270708
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
20
- Location
-
Chr.1:222676193 on Build GRCh38
- Set Membership
-
JSNP
- Context Sequence [VIC/FAM]
- TAGTGAGTAATGTCATTCCTGGTTC[C/T]GATGGCAACCTTGGTAATAAAGTAC
- Phenotype
-
MIM: 612375
MIM: 613455
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AIDA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147341411] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AIDA
- Gene Name
- axin interactor, dorsalization associated
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022831.2 |
|
Intron |
|
|
NP_073742.2 |
- Gene
- MIA3
- Gene Name
- MIA family member 3, ER export factor
There are no transcripts associated with this gene.
View Full Product Details