Product Details

SNP ID
rs17163416
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:222681788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGGGATGTGGACTGTGCAAGC[A/G]AGTCCTCTGTCCATGCTGGCCATGT
Phenotype
MIM: 612375
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AIDA PubMed Links

Gene Details

Gene
AIDA
Gene Name
axin interactor, dorsalization associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022831.2 Intron NP_073742.2

View Full Product Details