Product Details

SNP ID
rs35556780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:102190178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACATGCCCCTCAAAGGTAGCTGT[C/T]TATTCAGGTCAAGCCCATCTTCTCA
Phenotype
MIM: 604512
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL1RL2 PubMed Links

Gene Details

Gene
IL1RL2
Gene Name
interleukin 1 receptor like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003854.2 Intron NP_003845.2
XM_005264035.1 Intron XP_005264092.1
XM_005264036.1 Intron XP_005264093.1
XM_006712820.2 Intron XP_006712883.1
XM_006712822.2 Intron XP_006712885.1
XM_011512091.1 Intron XP_011510393.1
XM_011512092.1 Intron XP_011510394.1
XM_011512093.1 Intron XP_011510395.1
XM_011512094.1 Intron XP_011510396.1
XM_011512096.1 Intron XP_011510398.1
XM_017005174.1 Intron XP_016860663.1
XM_017005175.1 Intron XP_016860664.1
XM_017005176.1 Intron XP_016860665.1
XM_017005177.1 Intron XP_016860666.1
XM_017005178.1 Intron XP_016860667.1
XM_017005179.1 Intron XP_016860668.1
XM_017005180.1 Intron XP_016860669.1

View Full Product Details