Product Details

SNP ID
rs35786746
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177036550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTGCATCTCTTTTGTGTGTGTA[C/T]GCAGCCTCAGGGTGGCCATGAGTAT
Phenotype
MIM: 605308
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF346 PubMed Links

Gene Details

Gene
ZNF346
Gene Name
zinc finger protein 346
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308213.1 Intron NP_001295142.1
NM_001308214.1 Intron NP_001295143.1
NM_001308215.1 Intron NP_001295144.1
NM_001308216.1 Intron NP_001295145.1
NM_001308218.1 Intron NP_001295147.1
NM_001308219.1 Intron NP_001295148.1
NM_001308221.1 Intron NP_001295150.1
NM_001308223.1 Intron NP_001295152.1
NM_012279.3 Intron NP_036411.1
XM_005265860.2 Intron XP_005265917.1
XM_005265864.3 Intron XP_005265921.1
XM_011534493.1 Intron XP_011532795.1
XM_011534494.1 Intron XP_011532796.1
XM_011534495.2 Intron XP_011532797.1
XM_011534496.2 Intron XP_011532798.1
XM_011534497.2 Intron XP_011532799.1
XM_017009294.1 Intron XP_016864783.1
XM_017009295.1 Intron XP_016864784.1
XM_017009296.1 Intron XP_016864785.1
XM_017009297.1 Intron XP_016864786.1
XM_017009298.1 Intron XP_016864787.1
XM_017009299.1 Intron XP_016864788.1
XM_017009300.1 Intron XP_016864789.1
XM_017009301.1 Intron XP_016864790.1
XM_017009302.1 Intron XP_016864791.1
XM_017009303.1 Intron XP_016864792.1

View Full Product Details