Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001177316.1 | Intron | NP_001170787.1 | |||
NM_001177317.1 | Intron | NP_001170788.1 | |||
NM_080877.2 | Intron | NP_543153.1 | |||
XM_011518257.2 | Intron | XP_011516559.1 | |||
XM_011518258.1 | Intron | XP_011516560.1 | |||
XM_011518259.1 | Intron | XP_011516561.1 | |||
XM_011518260.1 | Intron | XP_011516562.1 | |||
XM_011518261.2 | Intron | XP_011516563.1 | |||
XM_017014290.1 | Intron | XP_016869779.1 | |||
XM_017014291.1 | Intron | XP_016869780.1 | |||
XM_017014292.1 | Intron | XP_016869781.1 |