Product Details

SNP ID
rs28509441
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27920300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCGCACCCGTTCCCTGGCGCCCT[A/G]GGCGCGCTGGAGCAGGGCAGCCCCC
Phenotype
MIM: 600733
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PDX1 PubMed Links

Gene Details

Gene
PDX1
Gene Name
pancreatic and duodenal homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000209.3 270 Silent Mutation CTA,CTG L54L NP_000200.1
Gene
PDX1-AS1
Gene Name
PDX1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details