Product Details

SNP ID
rs28631246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:143866724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTACTTCTTCATGTGGGTTGACAT[G/T]GGTATGCTTTATCAGTCTTGTTCCA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LTV1 PubMed Links

Gene Details

Gene
LTV1
Gene Name
LTV1 ribosome biogenesis factor
There are no transcripts associated with this gene.

Gene
ZC2HC1B
Gene Name
zinc finger C2HC-type containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013623.2 Intron NP_001013645.1

View Full Product Details