Product Details

SNP ID
rs28763268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149874441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTTGTCTTTCTGTGATATCAAT[G/T]GTGATGTCTTTCATTTATAATTTTA
Phenotype
MIM: 609243
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RAET1E PubMed Links

Gene Details

Gene
RAET1E
Gene Name
retinoic acid early transcript 1E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243325.1 Intron NP_001230254.1
NM_001243327.1 Intron NP_001230256.1
NM_001243328.1 Intron NP_001230257.1
NM_139165.2 Intron NP_631904.1
XM_011535475.2 Intron XP_011533777.1
XM_011535476.2 Intron XP_011533778.1
XM_011535478.2 Intron XP_011533780.1
XM_011535479.2 Intron XP_011533781.1
XM_017010285.1 Intron XP_016865774.1
XM_017010286.1 Intron XP_016865775.1
XM_017010287.1 Intron XP_016865776.1
XM_017010288.1 Intron XP_016865777.1
Gene
RAET1E-AS1
Gene Name
RAET1E antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details