Product Details

SNP ID
rs28477407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:54878183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTTCTGTCCAATGGAGTTCCCT[C/T]TGAACCCATAGTATAGGAATTAGTG
Phenotype
MIM: 600225
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GCH1 PubMed Links

Gene Details

Gene
GCH1
Gene Name
GTP cyclohydrolase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000161.2 Intron NP_000152.1
NM_001024024.1 Intron NP_001019195.1
NM_001024070.1 Intron NP_001019241.1
NM_001024071.1 Intron NP_001019242.1
XM_017021218.1 Intron XP_016876707.1
Gene
MIR4308
Gene Name
microRNA 4308
There are no transcripts associated with this gene.

View Full Product Details