Product Details

SNP ID
rs7081024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122562377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTTTCATGAAAGCAGCACCCCC[A/C]AACCCAGCCCTCCCACCCAGGTGGT
Phenotype
MIM: 601969
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DMBT1 PubMed Links

Gene Details

Gene
DMBT1
Gene Name
deleted in malignant brain tumors 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320644.1 Intron NP_001307573.1
NM_004406.2 Intron NP_004397.2
NM_007329.2 Intron NP_015568.2
NM_017579.2 Intron NP_060049.2
XM_006717660.3 Intron XP_006717723.1
XM_006717665.3 Intron XP_006717728.1
XM_011539388.2 Intron XP_011537690.1
XM_011539389.2 Intron XP_011537691.1
XM_011539390.2 Intron XP_011537692.1
XM_011539391.2 Intron XP_011537693.1
XM_011539392.2 Intron XP_011537694.1
XM_011539393.2 Intron XP_011537695.1
XM_011539394.2 Intron XP_011537696.1
XM_011539395.2 Intron XP_011537697.1
XM_011539396.2 Intron XP_011537698.1
XM_011539398.2 Intron XP_011537700.1
XM_011539399.2 Intron XP_011537701.1
XM_011539400.2 Intron XP_011537702.1
XM_011539401.2 Intron XP_011537703.1
XM_011539402.2 Intron XP_011537704.1
XM_011539403.2 Intron XP_011537705.1
XM_011539405.2 Intron XP_011537707.1
XM_011539407.2 Intron XP_011537709.1
XM_011539408.2 Intron XP_011537710.1
XM_011539409.2 Intron XP_011537711.1
XM_011539410.2 Intron XP_011537712.1
XM_011539411.2 Intron XP_011537713.1
XM_011539413.2 Intron XP_011537715.1
XM_011539414.2 Intron XP_011537716.1
XM_011539415.2 Intron XP_011537717.1
XM_017015798.1 Intron XP_016871287.1

View Full Product Details