Product Details

SNP ID
rs17886324
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:44800285 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGGAGTGGCCTCTCCTCGACTCG[C/T]TGCTAGGAAGGGGGCGGGACTCTCG
Phenotype
MIM: 607336 MIM: 602913 MIM: 611713
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BEST4 PubMed Links

Gene Details

Gene
BEST4
Gene Name
bestrophin 4
There are no transcripts associated with this gene.

Gene
BTBD19
Gene Name
BTB domain containing 19
There are no transcripts associated with this gene.

Gene
PLK3
Gene Name
polo like kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004073.3 61 UTR 5 NP_004064.2
Gene
TCTEX1D4
Gene Name
Tctex1 domain containing 4
There are no transcripts associated with this gene.

View Full Product Details