Product Details

SNP ID
rs17880618
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:44801586 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGATGGAGGGGGAGGGGGAGGGA[A/G]GGCTCACCAGGGGCTGAGGCAGTGG
Phenotype
MIM: 607336 MIM: 602913 MIM: 611713
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BEST4 PubMed Links

Gene Details

Gene
BEST4
Gene Name
bestrophin 4
There are no transcripts associated with this gene.

Gene
BTBD19
Gene Name
BTB domain containing 19
There are no transcripts associated with this gene.

Gene
PLK3
Gene Name
polo like kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004073.3 Intron NP_004064.2
Gene
TCTEX1D4
Gene Name
Tctex1 domain containing 4
There are no transcripts associated with this gene.

View Full Product Details