Product Details

SNP ID
rs17885815
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:44806156 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGCGGCCCCAGCACCACACTGCA[C/T]ACCAGCTTGTAGCGTGGCGGGCTGA
Phenotype
MIM: 602913 MIM: 611713
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTBD19 PubMed Links

Gene Details

Gene
BTBD19
Gene Name
BTB domain containing 19
There are no transcripts associated with this gene.

Gene
PLK3
Gene Name
polo like kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004073.3 656 Intron NP_004064.2
Gene
TCTEX1D4
Gene Name
Tctex1 domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013632.3 656 Silent Mutation GTA,GTG V171V NP_001013654.1
XM_006710614.3 656 Silent Mutation GTA,GTG V229V XP_006710677.1
XM_011541375.2 656 Silent Mutation GTA,GTG V171V XP_011539677.1
XM_011541376.2 656 Silent Mutation GTA,GTG V171V XP_011539678.1
XM_017001187.1 656 Silent Mutation GTA,GTG V212V XP_016856676.1

View Full Product Details