Product Details

SNP ID
rs28363197
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:46251331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTCCCCAAAACAACAACAAAAAA[A/G]CAACAGTTTTGTGTAATACTTGATA
Phenotype
MIM: 603615
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAD54L PubMed Links

Gene Details

Gene
RAD54L
Gene Name
RAD54-like (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142548.1 Intron NP_001136020.1
NM_003579.3 Intron NP_003570.2
XM_006710975.3 Intron XP_006711038.1
XM_011542299.2 Intron XP_011540601.1
XM_011542300.2 Intron XP_011540602.1

View Full Product Details