Product Details

SNP ID
rs28363251
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:46279578 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCTCAGGCAGAAATTGGGCTGGG[C/T]GCTTCTCAAACAGACGGCAGAAGGA
Phenotype
MIM: 603615
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC41 PubMed Links

Gene Details

Gene
LRRC41
Gene Name
leucine rich repeat containing 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006369.4 2101 Missense Mutation CAC,CGC H686R NP_006360.3
XM_017000074.1 2101 Missense Mutation CAC,CGC H694R XP_016855563.1
Gene
RAD54L
Gene Name
RAD54-like (S. cerevisiae)
There are no transcripts associated with this gene.

View Full Product Details