Product Details
- SNP ID
-
rs28605429
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:32234443 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGCAGAGGAAGGTGCCAAAAGAGG[A/G]GGCATAGGCGTGCCCAAACAGCGCC
- Phenotype
-
MIM: 603911
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EIF3I
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116838202] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EIF3I
- Gene Name
- eukaryotic translation initiation factor 3 subunit I
There are no transcripts associated with this gene.
- Gene
- MTMR9LP
- Gene Name
- myotubularin related protein 9-like, pseudogene
There are no transcripts associated with this gene.
View Full Product Details