Product Details

SNP ID
rs1137693
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:35931422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTCGAAGCACTCCCCCCAGCTC[A/C]ATGAATGGAAATCGGCTCCGCAGGT
Phenotype
MIM: 606228 MIM: 607355
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AGO1 PubMed Links

Gene Details

Gene
AGO1
Gene Name
argonaute 1, RISC catalytic component
There are no transcripts associated with this gene.

Gene
AGO3
Gene Name
argonaute 3, RISC catalytic component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024852.3 5811 UTR 5 NP_079128.2
NM_177422.2 5811 UTR 5 NP_803171.1
XM_005270575.3 5811 UTR 5 XP_005270632.1
XM_005270576.3 5811 Intron XP_005270633.1
XM_011540879.2 5811 Intron XP_011539181.1
XM_011540882.2 5811 Intron XP_011539184.1
XM_017000523.1 5811 UTR 5 XP_016856012.1
XM_017000524.1 5811 Intron XP_016856013.1
XM_017000525.1 5811 Intron XP_016856014.1
XM_017000526.1 5811 Intron XP_016856015.1
XM_017000527.1 5811 Intron XP_016856016.1
XM_017000528.1 5811 Intron XP_016856017.1

View Full Product Details