Product Details
- SNP ID
-
rs528520
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:57934048 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGATCAAGCAATCCTCCTGCCTCA[C/G]CTTTCCAAGTAGCTGGGACTATAAG
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GUSBP4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs190314253] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GUSBP4
- Gene Name
- glucuronidase, beta pseudogene 4
There are no transcripts associated with this gene.
- Gene
- LINC00680-GUSBP4
- Gene Name
- LINC00680-GUSBP4 readthrough
There are no transcripts associated with this gene.
View Full Product Details