Product Details

SNP ID
rs11550161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65855956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCAGAAGATAAACACCAGATCC[G/T]CCTTCTTGCTCTCCTTGGTCTCATA
Phenotype
MIM: 601442 MIM: 606591
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CFL1 PubMed Links

Gene Details

Gene
CFL1
Gene Name
cofilin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005507.2 524 Missense Mutation GAG,GCG E97A NP_005498.1
Gene
MUS81
Gene Name
MUS81 structure-specific endonuclease subunit
There are no transcripts associated with this gene.

Gene
SNX32
Gene Name
sorting nexin 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152760.2 524 Intron NP_689973.2

View Full Product Details