Product Details

SNP ID
rs28517596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:133980018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTAATATCACTGATTTGCCTTAA[G/T]TTTAAGAAGTTACTTCTTTAGTGAC
Phenotype
MIM: 604492
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC105379183 PubMed Links

Gene Details

Gene
LOC105379183
Gene Name
uncharacterized LOC105379183
There are no transcripts associated with this gene.

Gene
VDAC1
Gene Name
voltage dependent anion channel 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003374.2 Intron NP_003365.1
XM_005272075.3 Intron XP_005272132.1
XM_017009821.1 Intron XP_016865310.1
XM_017009822.1 Intron XP_016865311.1
XM_017009823.1 Intron XP_016865312.1

View Full Product Details