Product Details

SNP ID
rs28875781
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:83467163 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTATGTCTATTTCTGAAAGTTA[C/T]ATAGTTTTATGTTTTACATTTTAGT
Phenotype
MIM: 603362
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SH3GL3 PubMed Links

Gene Details

Gene
SH3GL3
Gene Name
SH3 domain containing GRB2 like 3, endophilin A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301108.1 Intron NP_001288037.1
NM_001301109.1 Intron NP_001288038.1
NM_001324182.1 Intron NP_001311111.1
NM_001324183.1 Intron NP_001311112.1
NM_001324184.1 Intron NP_001311113.1
NM_001324185.1 Intron NP_001311114.1
NM_001324186.1 Intron NP_001311115.1
NM_001324187.1 Intron NP_001311116.1
NM_003027.4 Intron NP_003018.3
XM_011521889.1 Intron XP_011520191.1
XM_011521891.2 Intron XP_011520193.1
XM_011521892.2 Intron XP_011520194.1
XM_017022486.1 Intron XP_016877975.1

View Full Product Details