Product Details
- SNP ID
-
hCV64435650
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:49055853 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATTTGGTACCTGAATTCTGAGAGG[A/G]AGAGTGCCCGAAGTTCAGACAGTGA
- Phenotype
-
MIM: 300947
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC120
PubMed Links
Gene Details
- Gene
- CCDC120
- Gene Name
- coiled-coil domain containing 120
There are no transcripts associated with this gene.
View Full Product Details