Product Details

SNP ID
rs61739693
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:158479959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGATCTTGGCAGAAAACCTCACC[A/G]TGGTCACCGAATTCCTGTTGCTGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10R2 PubMed Links

Gene Details

Gene
OR10R2
Gene Name
olfactory receptor family 10 subfamily R member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004472.1 82 Missense Mutation ATG,GTG M28V NP_001004472.1

View Full Product Details