Product Details
- SNP ID
-
rs61737506
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:174398334 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGTGGCATTACCTCCAGCAACAGT[C/T]GATAACAGGATTATTTTTGGAAAAA
- Phenotype
-
MIM: 605228
MIM: 614967
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CIR1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10497410] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CIR1
- Gene Name
- corepressor interacting with RBPJ, 1
There are no transcripts associated with this gene.
- Gene
- SCRN3
- Gene Name
- secernin 3
View Full Product Details