Product Details

SNP ID
rs61732026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:232456379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAGAACCCGGCCTTCTGGAACC[G/T]CCAGGCAGCTGAGGCCCTGGATGCT
Phenotype
MIM: 171740
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ALPI PubMed Links

Gene Details

Gene
ALPI
Gene Name
alkaline phosphatase, intestinal
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001631.4 186 Missense Mutation CGC,CTC R33L NP_001622.2

View Full Product Details