Product Details

SNP ID
rs61733452
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:148994248 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCACCAGGAGGCTGGTCGTGCT[C/T]GCCACCCCTCAGGTCTCAGACTCCA
Phenotype
MIM: 603942
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GYG1 PubMed Links

Gene Details

Gene
GYG1
Gene Name
glycogenin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184720.1 347 Silent Mutation CTC,CTT L38L NP_001171649.1
NM_001184721.1 347 Silent Mutation CTC,CTT L38L NP_001171650.1
NM_004130.3 347 Silent Mutation CTC,CTT L38L NP_004121.2
XM_017006275.1 347 Intron XP_016861764.1
XM_017006276.1 347 Intron XP_016861765.1

View Full Product Details