Product Details

SNP ID
rs61743521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:25312898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCGGGAATGGAGGTGGTGCTTC[C/T]TTTGGATCCTGCCGTCCCCGCCCCG
Phenotype
MIM: 611792
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZCCHC4 PubMed Links

Gene Details

Gene
ZCCHC4
Gene Name
zinc finger CCHC-type containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318148.1 123 Missense Mutation CCT,CTT P30L NP_001305077.1
NM_024936.2 123 Missense Mutation CCT,CTT P30L NP_079212.2
XM_011513835.1 123 Missense Mutation CCT,CTT P30L XP_011512137.1
XM_017008128.1 123 Missense Mutation CTT,TTT L25F XP_016863617.1
XM_017008129.1 123 Missense Mutation CCT,CTT P30L XP_016863618.1

View Full Product Details