Product Details

SNP ID
rs75657002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104598497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCTTCTCAGTAGGTGTTTTACT[G/A]CCTCCTTCACATCCTTGTTTCTAAG
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
OR13C2 PubMed Links

Gene Details

Gene
OR13C2
Gene Name
olfactory receptor family 13 subfamily C member 2
There are no transcripts associated with this gene.

Gene
OR13C5
Gene Name
olfactory receptor family 13 subfamily C member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004482.1 917 Missense Mutation GCA,GTA A306V NP_001004482.1

View Full Product Details