Product Details

SNP ID
rs61740767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:39276988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGATTCTAATCTTTATGGTTTT[C/T]CATGGGAATTGGTGATATGTGCAGC
Phenotype
MIM: 602132
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTAGE5 PubMed Links

Gene Details

Gene
CTAGE5
Gene Name
CTAGE family member 5, ER export factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001247988.1 454 Missense Mutation CCA,TCA P11S NP_001234917.1
NM_001247989.1 454 Missense Mutation CCA,TCA P40S NP_001234918.1
NM_001247990.1 454 Intron NP_001234919.1
NM_005930.3 454 Missense Mutation CCA,TCA P40S NP_005921.2
NM_203354.2 454 Missense Mutation CCA,TCA P28S NP_976229.1
NM_203355.2 454 Missense Mutation CCA,TCA P40S NP_976230.1
NM_203356.2 454 Missense Mutation CCA,TCA P11S NP_976231.1
XM_005267648.4 454 Intron XP_005267705.1
XM_006720148.2 454 Intron XP_006720211.1
XM_011536775.2 454 Intron XP_011535077.1
XM_011536778.2 454 Intron XP_011535080.1
XM_011536784.2 454 Intron XP_011535086.1
XM_011536785.2 454 Intron XP_011535087.1
XM_017021314.1 454 Intron XP_016876803.1
XM_017021315.1 454 Intron XP_016876804.1
XM_017021316.1 454 Intron XP_016876805.1
XM_017021317.1 454 Missense Mutation CCA,TCA P11S XP_016876806.1
XM_017021318.1 454 Intron XP_016876807.1
XM_017021319.1 454 Intron XP_016876808.1
XM_017021320.1 454 Intron XP_016876809.1
XM_017021321.1 454 Intron XP_016876810.1
XM_017021322.1 454 Intron XP_016876811.1
XM_017021323.1 454 Intron XP_016876812.1
XM_017021324.1 454 Intron XP_016876813.1
XM_017021325.1 454 Intron XP_016876814.1
XM_017021326.1 454 Intron XP_016876815.1
XM_017021327.1 454 Intron XP_016876816.1
XM_017021328.1 454 Intron XP_016876817.1
XM_017021329.1 454 Intron XP_016876818.1
XM_017021330.1 454 Intron XP_016876819.1
Gene
LOC100288846
Gene Name
uncharacterized LOC100288846
There are no transcripts associated with this gene.

View Full Product Details