Product Details
- SNP ID
-
rs17424311
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:1636691 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGGGCCAGACCCACCTTGAACAC[T/C]TTGTTGATCTGATCGATTTCTGACT
- Phenotype
-
MIM: 176873
MIM: 611141
MIM: 603321
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDK11B
PubMed Links
Gene Details
- Gene
- CDK11B
- Gene Name
- cyclin dependent kinase 11B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001291345.1 |
1845 |
Silent Mutation |
AAA,AAG |
K626K |
NP_001278274.1 |
NM_001787.2 |
1845 |
Silent Mutation |
AAA,AAG |
K649K |
NP_001778.2 |
NM_033486.2 |
1845 |
Silent Mutation |
AAA,AAG |
K636K |
NP_277021.2 |
NM_033487.2 |
1845 |
Silent Mutation |
AAA,AAG |
K380K |
NP_277022.1 |
NM_033489.2 |
1845 |
Silent Mutation |
AAA,AAG |
K602K |
NP_277024.2 |
NM_033490.2 |
1845 |
Silent Mutation |
AAA,AAG |
K419K |
NP_277025.1 |
XM_006711065.3 |
1845 |
Silent Mutation |
AAA,AAG |
K639K |
XP_006711128.1 |
XM_011542490.2 |
1845 |
Silent Mutation |
AAA,AAG |
K658K |
XP_011540792.1 |
XM_011542491.2 |
1845 |
Silent Mutation |
AAA,AAG |
K658K |
XP_011540793.1 |
XM_011542492.2 |
1845 |
Silent Mutation |
AAA,AAG |
K658K |
XP_011540794.1 |
XM_011542493.2 |
1845 |
Silent Mutation |
AAA,AAG |
K648K |
XP_011540795.1 |
XM_011542494.2 |
1845 |
Silent Mutation |
AAA,AAG |
K624K |
XP_011540796.1 |
XM_011542495.2 |
1845 |
Silent Mutation |
AAA,AAG |
K614K |
XP_011540797.1 |
XM_011542496.2 |
1845 |
Silent Mutation |
AAA,AAG |
K432K |
XP_011540798.1 |
XM_017002925.1 |
1845 |
Silent Mutation |
AAA,AAG |
K645K |
XP_016858414.1 |
XM_017002926.1 |
1845 |
Silent Mutation |
AAA,AAG |
K635K |
XP_016858415.1 |
XM_017002927.1 |
1845 |
Silent Mutation |
AAA,AAG |
K624K |
XP_016858416.1 |
XM_017002928.1 |
1845 |
Silent Mutation |
AAA,AAG |
K615K |
XP_016858417.1 |
- Gene
- MIB2
- Gene Name
- mindbomb E3 ubiquitin protein ligase 2
There are no transcripts associated with this gene.
- Gene
- MMP23B
- Gene Name
- matrix metallopeptidase 23B
There are no transcripts associated with this gene.
View Full Product Details