Product Details

SNP ID
rs28385651
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:27369839 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTGGCCAGGAAAGGTGGTTCCC[C/T]GAGGCACTCCCTCATAACCACTTCC
Phenotype
MIM: 604973 MIM: 604468
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD164L2 PubMed Links

Gene Details

Gene
CD164L2
Gene Name
CD164 molecule like 2
There are no transcripts associated with this gene.

Gene
FCN3
Gene Name
ficolin 3
There are no transcripts associated with this gene.

Gene
MAP3K6
Gene Name
mitogen-activated protein kinase kinase kinase 6
There are no transcripts associated with this gene.

View Full Product Details