Product Details

SNP ID
rs41267843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:161132519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCTCATGCCCTCACCTAGAAA[C/T]GCCTTCTCGGCTGAATGGCTGTTGG
Phenotype
MIM: 614795
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGPAT4 PubMed Links

Gene Details

Gene
AGPAT4
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020133.2 5385 UTR 3 NP_064518.1
XM_005267053.3 5385 UTR 3 XP_005267110.1
XM_006715514.3 5385 UTR 3 XP_006715577.1
XM_017011059.1 5385 UTR 3 XP_016866548.1

View Full Product Details