Product Details

SNP ID
hCV86387920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44780911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTGCCCATCCCCACAGGCGACA[C/G]CTGGCCAAGCTGGCCATCATCTTCA
Phenotype
MIM: 608453
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CBLC PubMed Links

Gene Details

Gene
CBLC
Gene Name
Cbl proto-oncogene C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130852.1 423 Missense Mutation CAC,CAG H120Q NP_001124324.1
NM_012116.3 423 Missense Mutation CAC,CAG H120Q NP_036248.3

View Full Product Details