Product Details

SNP ID
rs41304733
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:35802419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTTTCTGCTCCTCTTCCATTGCC[A/G]TCACAACCACCTCATCAAGTGAATC
Phenotype
MIM: 300762
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAGEB16 PubMed Links

Gene Details

Gene
MAGEB16
Gene Name
MAGE family member B16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099921.1 391 Missense Mutation ATC,GTC I75V NP_001093391.1
XM_011545453.2 391 Missense Mutation ATC,GTC I75V XP_011543755.1
XM_011545454.2 391 Missense Mutation ATC,GTC I75V XP_011543756.1

View Full Product Details