Product Details

SNP ID
rs41301495
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:130495246 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAAATGGAGAAGTTCTGTGGGTG[C/T]GGTGTTATCCTTCCACGACAGCCAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM45BP PubMed Links

Gene Details

Gene
FAM45BP
Gene Name
family with sequence similarity 45, member A pseudogene
There are no transcripts associated with this gene.

View Full Product Details