Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001184977.1 | 1643 | UTR 3 | NP_001171906.1 | ||
NM_018244.4 | 1643 | UTR 3 | NP_060714.3 | ||
NM_199487.2 | 1643 | UTR 3 | NP_955781.2 | ||
XM_011528877.1 | 1643 | UTR 3 | XP_011527179.1 | ||
XM_011528878.1 | 1643 | UTR 3 | XP_011527180.1 | ||
XM_011528879.1 | 1643 | UTR 3 | XP_011527181.1 | ||
XM_011528880.1 | 1643 | UTR 3 | XP_011527182.1 | ||
XM_011528881.2 | 1643 | UTR 3 | XP_011527183.1 | ||
XM_011528882.1 | 1643 | UTR 3 | XP_011527184.1 | ||
XM_011528883.1 | 1643 | UTR 3 | XP_011527185.1 | ||
XM_011528884.1 | 1643 | UTR 3 | XP_011527186.1 |