Product Details

SNP ID
rs41285732
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:99712167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCCAGTTAAACATTTGCCTTCTA[C/T]CAAAATAAGAACCAGTCAGTATTCT
Phenotype
MIM: 611578
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FRRS1 PubMed Links

Gene Details

Gene
FRRS1
Gene Name
ferric chelate reductase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013660.2 Intron NP_001013682.2
XM_005270861.3 Intron XP_005270918.1
XM_011541451.2 Intron XP_011539753.1
XM_011541452.1 Intron XP_011539754.1
XM_017001271.1 Intron XP_016856760.1

View Full Product Details