Product Details

SNP ID
rs41306540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:125766721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCAAACCACTTCTCTGAAATAA[A/G]CTTTTTTGGAAATAAGCCATTAGCA
Phenotype
MIM: 608416
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EDRF1 PubMed Links

Gene Details

Gene
EDRF1
Gene Name
erythroid differentiation regulatory factor 1
There are no transcripts associated with this gene.

Gene
MMP21
Gene Name
matrix metallopeptidase 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_147191.1 1651 Missense Mutation CTT,TTT L551F NP_671724.1

View Full Product Details