Product Details

SNP ID
rs57740496
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:173717699 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCTTTTAGTCTCATGTCTTCCT[A/G]TATTTGCATCACTAGCACCTAACGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHL20 PubMed Links

Gene Details

Gene
KLHL20
Gene Name
kelch like family member 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014458.3 Intron NP_055273.2
XM_005245093.3 Intron XP_005245150.1
XM_011509420.2 Intron XP_011507722.1
XM_017001052.1 Intron XP_016856541.1
XM_017001053.1 Intron XP_016856542.1
XM_017001054.1 Intron XP_016856543.1

View Full Product Details