Product Details

SNP ID
rs62541897
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:20114497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTGTTGAGCACCTACCGTGTCAC[A/G]GCAGGAGAGGACTGAACGATAGGGA
Phenotype
MIM: 605630
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLA2G2D PubMed Links

Gene Details

Gene
PLA2G2D
Gene Name
phospholipase A2 group IID
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271814.1 Intron NP_001258743.1
NM_012400.3 Intron NP_036532.1

View Full Product Details