Product Details

SNP ID
hCV88397953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:16047922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCAAGGTTCTGGAATCCCGGAG[C/G]TGAAGACCATGTTGGCGGGTGTGGT
Phenotype
MIM: 602023
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CLCNKB PubMed Links

Gene Details

Gene
CLCNKB
Gene Name
chloride voltage-gated channel Kb
There are no transcripts associated with this gene.

Gene
FAM131C
Gene Name
family with sequence similarity 131 member C
There are no transcripts associated with this gene.

View Full Product Details