Product Details

SNP ID
rs60399987
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:165873530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGAGTGAAAGCTTCTATGCCAG[A/G]GTGCTGTTAAACAGAGCTGGGTAAA
Phenotype
MIM: 612014
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100506124 PubMed Links

Gene Details

Gene
LOC100506124
Gene Name
uncharacterized LOC100506124
There are no transcripts associated with this gene.

Gene
TTC21B
Gene Name
tetratricopeptide repeat domain 21B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024753.4 5283 UTR 3 NP_079029.3
XM_006712761.1 5283 Intron XP_006712824.1
XM_011511871.2 5283 UTR 3 XP_011510173.1
XM_011511872.2 5283 UTR 3 XP_011510174.1
XM_017004967.1 5283 Intron XP_016860456.1
XM_017004968.1 5283 UTR 3 XP_016860457.1
XM_017004969.1 5283 UTR 3 XP_016860458.1

View Full Product Details