Product Details

SNP ID
rs58846152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:188734646 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGTTTAATATTTTCCACCCCCTC[A/G]ATGAGTTAAGACACTTTAGCCTCAC
Phenotype
MIM: 606423
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DIRC1 PubMed Links

Gene Details

Gene
DIRC1
Gene Name
disrupted in renal carcinoma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052952.2 546 Missense Mutation TCG,TTG S92L NP_443184.1
XM_017003281.1 546 Missense Mutation TCG,TTG S92L XP_016858770.1

View Full Product Details